siRNA Details
| Gene Symbol: LOC101927869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109963.1 | 4 | 835 | |||
| GenBank mRNA | BC043533.1 | - | 4 | 835 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927869| Gene Symbol: LOC101927869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109963.1 | 4 | 835 | |||
| GenBank mRNA | BC043533.1 | - | 4 | 835 | ||