siRNA Details
| Gene Symbol: LOC729348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130931.1 | 3 | 311 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE884391.1 | - | 3 | 301 | Chr. 2 - 66383316 - 66391355 [+] on Build GRCh38 | |

LOC729348
Genomic Map