siRNA Details
Gene Symbol: LVCAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131961.1 | 4 | 1441 | |||
GenBank mRNA | CB751600.1 | - | 4 | 1420 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LVCAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131961.1 | 4 | 1441 | |||
GenBank mRNA | CB751600.1 | - | 4 | 1420 |