siRNA Details
Gene Symbol: LOC101928111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135575.1 | 2 | 343 | |||
NR_135576.1 | 2 | 492 | ||||
NR_135577.1 | 2 | 375 | ||||
GenBank mRNA | BM704421.1 | - | 2 | 375 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135575.1 | 2 | 343 | |||
NR_135576.1 | 2 | 492 | ||||
NR_135577.1 | 2 | 375 | ||||
GenBank mRNA | BM704421.1 | - | 2 | 375 |