siRNA Details
Gene Symbol: KCNIP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045118.1 | 3 | 724 | |||
GenBank mRNA | BX090826.1 | - | 3 | 662 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI827949.1 | - | 3 | 713 | Chr. 10 - 101819078 - 101828772 [+] on Build GRCh38 |
