siRNA Details
| Gene Symbol: LOC100507670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135595.1 | 2 | 545 | |||
| GenBank mRNA | BC021024.1 | - | 2 | 545 | ||
| BE385446.1 | - | 2 | 573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507670| Gene Symbol: LOC100507670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135595.1 | 2 | 545 | |||
| GenBank mRNA | BC021024.1 | - | 2 | 545 | ||
| BE385446.1 | - | 2 | 573 | |||