siRNA Details
| Gene Symbol: LOC100507670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135595.1 | 1 | 376 | |||
| GenBank mRNA | DA674483.1 | - | 1 | 344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507670| Gene Symbol: LOC100507670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135595.1 | 1 | 376 | |||
| GenBank mRNA | DA674483.1 | - | 1 | 344 | ||