siRNA Details
| Gene Symbol: ARHGEF26-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037901.1 | 4 | 1360 | |||
| GenBank mRNA | BC038431.1 | - | 4 | 1362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARHGEF26-AS1| Gene Symbol: ARHGEF26-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037901.1 | 4 | 1360 | |||
| GenBank mRNA | BC038431.1 | - | 4 | 1362 | ||