siRNA Details
| Gene Symbol: LOC101929662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120670.1 | 3 | 2099 | |||
| GenBank mRNA | BC041821.1 | - | 3 | 2094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929662| Gene Symbol: LOC101929662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120670.1 | 3 | 2099 | |||
| GenBank mRNA | BC041821.1 | - | 3 | 2094 | ||