siRNA Details
| Gene Symbol: WDR86-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034012.2 | 5 | 688 | |||
| NR_034013.2 | 4 | 603 | ||||
| GenBank mRNA | AK095986.1 | - | 4 | 528 | ||
| BC043579.1 | - | 4 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR86-AS1| Gene Symbol: WDR86-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034012.2 | 5 | 688 | |||
| NR_034013.2 | 4 | 603 | ||||
| GenBank mRNA | AK095986.1 | - | 4 | 528 | ||
| BC043579.1 | - | 4 | 479 | |||