siRNA Details
Gene Symbol: THRA1/BTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131239.1 | 3 | 421 | |||
NR_131240.1 | 2 | 361 | ||||
GenBank mRNA | BG185647.1 | - | 1 | 196 | ||
BG197541.1 | - | 1 | 204 | |||
DA697253.1 | - | 3 | 393 |
