siRNA Details
Gene Symbol: THRA1/BTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131239.1 | 1 | 68 | |||
NR_131240.1 | 1 | 68 | ||||
GenBank mRNA | DA697253.1 | - | 1 | 40 | ||
S71020.1 | - | 1 | 82 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: THRA1/BTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131239.1 | 1 | 68 | |||
NR_131240.1 | 1 | 68 | ||||
GenBank mRNA | DA697253.1 | - | 1 | 40 | ||
S71020.1 | - | 1 | 82 |