siRNA Details
| Gene Symbol: LOC101927577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110264.1 | 2 | 481 | |||
| NR_110265.1 | 2 | 195 | ||||
| GenBank mRNA | CD557966.1 | - | 2 | 481 | ||
| HY009512.1 | - | 2 | 184 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927577| Gene Symbol: LOC101927577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110264.1 | 2 | 481 | |||
| NR_110265.1 | 2 | 195 | ||||
| GenBank mRNA | CD557966.1 | - | 2 | 481 | ||
| HY009512.1 | - | 2 | 184 | |||