siRNA Details
| Gene Symbol: LOC101927577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110264.1 | 1 | 65 | |||
| NR_110265.1 | 1 | 65 | ||||
| GenBank mRNA | CD557966.1 | - | 1 | 65 | ||
| HY009512.1 | - | 1 | 54 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB448856.1 | - | 1 | 35 | Chr. 2 - 66697162 - 66703197 [-] on Build GRCh38 | |

LOC101927577
Genomic Map