siRNA Details
| Gene Symbol: LINC01056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033369.1 | 4, 5 | 527 | |||
| GenBank mRNA | BC038718.1 | - | 4, 5 | 527 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI460714.1 | - | 4, 5 | 531 | Chr. 20 - 63038011 - 63053124 [+] on Build GRCh38 | |

LINC01056
Genomic Map