siRNA Details
| Gene Symbol: LOC100506100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046240.1 | 2 | 377 | |||
| GenBank mRNA | BX090437.1 | - | 2 | 371 | ||
| EB386762.1 | - | 2 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506100| Gene Symbol: LOC100506100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046240.1 | 2 | 377 | |||
| GenBank mRNA | BX090437.1 | - | 2 | 371 | ||
| EB386762.1 | - | 2 | 377 | |||