siRNA Details
Gene Symbol: LOC101927766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109891.1 | 3 | 5419 | |||
GenBank mRNA | AL833484.1 | - | 3 | 5419 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109891.1 | 3 | 5419 | |||
GenBank mRNA | AL833484.1 | - | 3 | 5419 |