siRNA Details
| Gene Symbol: LOC63930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK310606.1 | - | 2 | 394 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB127133.1 | - | 2 | 394 | Chr. 20 - 63009383 - 63083660 [+] on Build GRCh38 | |

LOC63930
Genomic Map