siRNA Details
| Gene Symbol: LOC101929680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110260.1 | 12 | 1696 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929680| Gene Symbol: LOC101929680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110260.1 | 12 | 1696 | |||