siRNA Details
| Gene Symbol: LOC101927653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120452.1 | 8 | 1908 | |||
| GenBank mRNA | AK093124.1 | - | 8 | 1908 | ||
| BX109998.1 | - | 1 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927653| Gene Symbol: LOC101927653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120452.1 | 8 | 1908 | |||
| GenBank mRNA | AK093124.1 | - | 8 | 1908 | ||
| BX109998.1 | - | 1 | 406 | |||