siRNA Details
Gene Symbol: CTD-2270F17.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109899.1 | 2 | 297 | |||
GenBank mRNA | AK311026.1 | - | 2 | 298 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CTD-2270F17.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109899.1 | 2 | 297 | |||
GenBank mRNA | AK311026.1 | - | 2 | 298 |