siRNA Details
| Gene Symbol: CTD-2270F17.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109899.1 | 3 | 588 | |||
| GenBank mRNA | AK311026.1 | - | 3 | 589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTD-2270F17.1| Gene Symbol: CTD-2270F17.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109899.1 | 3 | 588 | |||
| GenBank mRNA | AK311026.1 | - | 3 | 589 | ||