siRNA Details
Gene Symbol: MBNL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027037.1 | 2 | 955 | |||
NR_027038.1 | 2 | 966 | ||||
GenBank mRNA | AK126417.1 | - | 1 | 52 | ||
AK304857.1 | - | 2 | 875 | |||
AK316515.1 | - | 2 | 895 |
