siRNA Details
| Gene Symbol: C9orf41-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121183.1 | 5 | 634 | |||
| GenBank mRNA | BC043649.2 | - | 5 | 630 | ||
| BI552900.1 | - | 5 | 636 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf41-AS1| Gene Symbol: C9orf41-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121183.1 | 5 | 634 | |||
| GenBank mRNA | BC043649.2 | - | 5 | 630 | ||
| BI552900.1 | - | 5 | 636 | |||