siRNA Details
| Gene Symbol: C9orf41-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121183.1 | 2 | 172 | |||
| GenBank mRNA | BC043649.2 | - | 2 | 168 | ||
| BI552900.1 | - | 2 | 173 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB465744.1 | - | 2 | 161 | Chr. 9 - 74952976 - 74991677 [+] on Build GRCh38 | |

C9orf41-AS1
Genomic Map