siRNA Details
| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 6 | 1359 | |||
| NR_027429.1 | 5 | 1272 | ||||
| NR_027430.1 | 5 | 1148 | ||||
| NR_027431.2 | 5 | 1122 | ||||
| NR_027432.1 | 4 | 1035 | ||||
| GenBank mRNA | AK021673.1 | - | 5 | 991 | ||
| AK096758.1 | - | 6 | 1061 | |||
| AK308811.1 | - | 4 | 588 | |||
| AK309654.1 | - | 5 | 771 | |||
| AK310182.1 | - | 5 | 824 | |||
| AK310261.1 | - | 4 | 725 | |||
| BC028046.1 | - | 4 | 696 | |||

TMEM254-AS1
Genomic Map