siRNA Details
| Gene Symbol: LCMT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120306.1 | 1 | 71 | |||
| GenBank mRNA | AL600125.1 | - | 1 | 72 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY060601.1 | - | 1 | 40 | Chr. 16 - 25103232 - 25111523 [-] on Build GRCh38 | |

LCMT1-AS1
Genomic Map