siRNA Details
| Gene Symbol: LINC00517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135286.1 | 2 | 2100 | |||
| GenBank mRNA | AK125955.1 | - | 2 | 2101 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00517| Gene Symbol: LINC00517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135286.1 | 2 | 2100 | |||
| GenBank mRNA | AK125955.1 | - | 2 | 2101 | ||