siRNA Details
| Gene Symbol: LOC102467147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104664.1 | 2 | 267 | |||
| GenBank mRNA | BX484633.1 | - | 2 | 268 | ||
| DB291781.1 | - | 2 | 469 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CV339640.1 | - | 2 | 182 | Chr. 5 - 56457854 - 56481673 [-] on Build GRCh38 | |

LOC102467147
Genomic Map