siRNA Details
| Gene Symbol: FOXP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126463.1 | 5 | 685 | |||
| GenBank mRNA | AA399395.1 | - | 2 | 302 | ||
| BC040624.1 | - | 5 | 674 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXP1-AS1| Gene Symbol: FOXP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126463.1 | 5 | 685 | |||
| GenBank mRNA | AA399395.1 | - | 2 | 302 | ||
| BC040624.1 | - | 5 | 674 | |||