siRNA Details
| Gene Symbol: TM4SF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046650.1 | 1 | 61 | |||
| NR_109809.1 | 1 | 61 | ||||
| NR_109810.1 | 1 | 61 | ||||
| GenBank mRNA | CB956551.1 | - | 1 | 64 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM4SF1-AS1| Gene Symbol: TM4SF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046650.1 | 1 | 61 | |||
| NR_109809.1 | 1 | 61 | ||||
| NR_109810.1 | 1 | 61 | ||||
| GenBank mRNA | CB956551.1 | - | 1 | 64 | ||