siRNA Details
| Gene Symbol: ZRANB2-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046217.1 | 4 | 735 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZRANB2-AS2| Gene Symbol: ZRANB2-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046217.1 | 4 | 735 | |||