siRNA Details
| Gene Symbol: LOC105369507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135102.1 | 2 | 332 | |||
| NR_135103.1 | 2 | 335 | ||||
| GenBank mRNA | BX106383.1 | - | 2 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369507| Gene Symbol: LOC105369507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135102.1 | 2 | 332 | |||
| NR_135103.1 | 2 | 335 | ||||
| GenBank mRNA | BX106383.1 | - | 2 | 332 | ||