siRNA Details
| Gene Symbol: LOC100506682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110024.1 | 4 | 525 | |||
| GenBank mRNA | BX095908.1 | - | 2 | 342 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506682| Gene Symbol: LOC100506682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110024.1 | 4 | 525 | |||
| GenBank mRNA | BX095908.1 | - | 2 | 342 | ||