siRNA Details
| Gene Symbol: LINC01256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126401.1 | 1, 2 | 197 | |||
| GenBank mRNA | BX088800.1 | - | 1, 2 | 197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01256| Gene Symbol: LINC01256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126401.1 | 1, 2 | 197 | |||
| GenBank mRNA | BX088800.1 | - | 1, 2 | 197 | ||