siRNA Details
| Gene Symbol: PCAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026555.1 | 2 | 371 | |||
| GenBank mRNA | AF388200.1 | - | 2 | 366 | ||
| AF462307.1 | - | 2 | 322 | |||
| AK056825.1 | - | 2 | 371 | |||
| BC105591.1 | - | 2 | 469 | |||
| BC125139.1 | - | 2 | 368 | |||

PCAT4
Genomic Map