siRNA Details
| Gene Symbol: LINC01333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105010.1 | 1 | 63 | |||
| GenBank mRNA | BC043537.1 | - | 1 | 63 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI828590.1 | - | 1 | 63 | Chr. 5 - 74322486 - 74328262 [+] on Build GRCh38 | |

LINC01333
Genomic Map