siRNA Details
| Gene Symbol: SMIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525928.1 | XP_011524230.1 | 3 | 962 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM21| Gene Symbol: SMIM21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525928.1 | XP_011524230.1 | 3 | 962 | ||