siRNA Details
| Gene Symbol: LOC101927768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125845.1 | 2, 3 | 712 | |||
| GenBank mRNA | BC042098.1 | - | 2, 3 | 712 | ||
| CD643945.1 | - | 2, 3 | 444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927768| Gene Symbol: LOC101927768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125845.1 | 2, 3 | 712 | |||
| GenBank mRNA | BC042098.1 | - | 2, 3 | 712 | ||
| CD643945.1 | - | 2, 3 | 444 | |||