siRNA Details
| Gene Symbol: LOC101927592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104646.1 | 5, 6 | 854 | |||
| GenBank mRNA | BC032874.2 | - | 5, 6 | 854 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927592| Gene Symbol: LOC101927592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104646.1 | 5, 6 | 854 | |||
| GenBank mRNA | BC032874.2 | - | 5, 6 | 854 | ||