siRNA Details
| Gene Symbol: DOCK4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103806.1 | 2 | 537 | |||
| GenBank mRNA | BC043243.2 | - | 2 | 506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DOCK4-AS1| Gene Symbol: DOCK4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103806.1 | 2 | 537 | |||
| GenBank mRNA | BC043243.2 | - | 2 | 506 | ||