siRNA Details
Gene Symbol: LOC254028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122043.1 | 2 | 300 | |||
GenBank mRNA | AK093210.1 | - | 1 | 240 | ||
BI830942.1 | - | 2 | 300 | |||
DB034207.1 | - | 2 | 244 | |||
DB053670.1 | - | 2 | 371 |
