siRNA Details
Gene Symbol: LOC254028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122043.1 | 5 | 1895 | |||
GenBank mRNA | AK093210.1 | - | 4 | 1840 | ||
BX247965.1 | - | 3 | 1401 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC254028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122043.1 | 5 | 1895 | |||
GenBank mRNA | AK093210.1 | - | 4 | 1840 | ||
BX247965.1 | - | 3 | 1401 |