siRNA Details
| Gene Symbol: LOC101929454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109944.1 | 3 | 441 | |||
| GenBank mRNA | BC002741.2 | - | 3 | 441 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE394211.1 | - | 3 | 422 | Chr. 5 - 15192139 - 15266541 [-] on Build GRCh38 | |

LOC101929454
Genomic Map