siRNA Details
| Gene Symbol: LOC100506498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135238.1 | 2 | 405 | |||
| GenBank mRNA | BC013914.1 | - | 2 | 350 | ||
| BC013921.1 | - | 2 | 350 | |||
| BC038204.1 | - | 2 | 405 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506498| Gene Symbol: LOC100506498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135238.1 | 2 | 405 | |||
| GenBank mRNA | BC013914.1 | - | 2 | 350 | ||
| BC013921.1 | - | 2 | 350 | |||
| BC038204.1 | - | 2 | 405 | |||