siRNA Details
Gene Symbol: SRP14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040059.1 | 4 | 794 | |||
NR_040060.1 | 3 | 511 | ||||
GenBank mRNA | AK127696.1 | - | 3 | 509 | ||
EB388563.1 | - | 4 | 718 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SRP14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040059.1 | 4 | 794 | |||
NR_040060.1 | 3 | 511 | ||||
GenBank mRNA | AK127696.1 | - | 3 | 509 | ||
EB388563.1 | - | 4 | 718 |