siRNA Details
| Gene Symbol: LCMT1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039998.1 | 2 | 614 | |||
| GenBank mRNA | AK056728.1 | - | 2 | 614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCMT1-AS2| Gene Symbol: LCMT1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039998.1 | 2 | 614 | |||
| GenBank mRNA | AK056728.1 | - | 2 | 614 | ||