siRNA Details
| Gene Symbol: LINC01583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120367.1 | 2 | 149 | |||
| GenBank mRNA | BC043257.2 | - | 2 | 149 | ||
| BX104219.1 | - | 2 | 112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01583| Gene Symbol: LINC01583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120367.1 | 2 | 149 | |||
| GenBank mRNA | BC043257.2 | - | 2 | 149 | ||
| BX104219.1 | - | 2 | 112 | |||