siRNA Details
| Gene Symbol: LINC01210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109987.1 | 4 | 424 | |||
| GenBank mRNA | BC038725.2 | - | 3 | 204 | ||
| BI460052.1 | - | 4 | 239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01210| Gene Symbol: LINC01210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109987.1 | 4 | 424 | |||
| GenBank mRNA | BC038725.2 | - | 3 | 204 | ||
| BI460052.1 | - | 4 | 239 | |||