siRNA Details
| Gene Symbol: TMEM246-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121573.1 | 4 | 547 | |||
| NR_121574.1 | 3 | 509 | ||||
| NR_121575.1 | 2 | 425 | ||||
| NR_121576.1 | 3 | 380 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM246-AS1| Gene Symbol: TMEM246-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121573.1 | 4 | 547 | |||
| NR_121574.1 | 3 | 509 | ||||
| NR_121575.1 | 2 | 425 | ||||
| NR_121576.1 | 3 | 380 | ||||